
2-Aminobenzamide

Synonym(s):
Anthranilamide, 2-AB, 2-Aminobenzamide, Anthranilic acid amide
Linear Formula:
2-(H2N)C6H4CONH2
CAS Number:
Molecular Weight:
136.15
Products
Brand
Formula Weight
Melting Point (°C)
Format
Quality Segment
Markush Class
Markush Group
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing