
2-Amino-3-bromobenzoic acid

Empirical Formula (Hill Notation):
C7H6BrNO2
CAS Number:
Molecular Weight:
216.03
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing