
2,4-Dichloro-6,7-dimethoxyquinazoline

Empirical Formula (Hill Notation):
C10H8Cl2N2O2
CAS Number:
Molecular Weight:
259.09
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing