![2-[4-(2-Hydroxy-2-methylpropyl)-phenyl]propanoic Acid certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/structures/133/400/476867f7-e388-43a9-97a1-8b0458a8e4e3/640/476867f7-e388-43a9-97a1-8b0458a8e4e3.png)
2-[4-(2-Hydroxy-2-methylpropyl)-phenyl]propanoic Acid
![2-[4-(2-Hydroxy-2-methylpropyl)-phenyl]propanoic Acid certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/structures/133/400/476867f7-e388-43a9-97a1-8b0458a8e4e3/640/476867f7-e388-43a9-97a1-8b0458a8e4e3.png)
Synonym(s):
2-Hydroxyibuprofen, 2-[4-(2-Hydroxy-2-methylpropyl)phenyl]propionic acid
Empirical Formula (Hill Notation):
C13H18O3
CAS Number:
Molecular Weight:
222.28
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing