Skip to Content
Merck
2-(3-Bromopropoxy)tetrahydro-2H-pyran 98%

2-(3-Bromopropoxy)tetrahydro-2H-pyran

2-(3-Bromopropoxy)tetrahydro-2H-pyran 98%
Synonym(s):
1-Bromo-3-(tetrahydropyranyloxy)propane
Empirical Formula (Hill Notation):
C8H15BrO2
CAS Number:
Molecular Weight:
223.11

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing