
2,3,5,4′-Tetrahydroxystilbene 2-O-β-D-glucoside

Synonym(s):
2,4-Dihydroxy-6-[(1E)-2-(4-hydroxyphenyl)ethenyl]phenyl β-D-Glucopyranoside, EH-201, THSG, Tetrahydroxystilbene glucoside
Empirical Formula (Hill Notation):
C20H22O9
CAS Number:
Molecular Weight:
406.38
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing