Skip to Content
Merck
2,2,5,7,8-Pentamethyl-chromane-6-sulfonyl chloride ≥97.0% (HPLC)

2,2,5,7,8-Pentamethyl-chromane-6-sulfonyl chloride

2,2,5,7,8-Pentamethyl-chromane-6-sulfonyl chloride ≥97.0% (HPLC)
Synonym(s):
3,4-Dihydro-2,2,5,7,8-pentamethyl-2H-1-benzopyran-6-sulfonyl chloride, Pmc-Cl
Empirical Formula (Hill Notation):
C14H19ClO3S
CAS Number:
Molecular Weight:
302.82

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing