Skip to Content
Merck
1-Amino-1-cyclobutanecarboxylic acid 97%

1-Amino-1-cyclobutanecarboxylic acid

1-Amino-1-cyclobutanecarboxylic acid 97%
Linear Formula:
H2NC4H6CO2H
CAS Number:
Molecular Weight:
115.13

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing