
1-(2,4-Dichlorobenzoylmethyl)imidazole

Empirical Formula (Hill Notation):
C11H8Cl2N2O
CAS Number:
Molecular Weight:
255.10
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing