Skip to Content
Merck
All Photos(1)

Documents

GERPN4401

Amdex Streptavidin Horseradish Peroxidase Conjugate

Cytiva RPN4401

Synonym(s):

HRP streptavidin, Streptavidin HRP conjugate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

form

liquid

packaging

pkg of 0.5 mL

manufacturer/tradename

Cytiva RPN4401

storage temp.

2-8°C

General description

Amdex high-performance conjugates offer a novel approach to obtaining significantly improved sensitivity for high-sensitivity assay development. Improvement is gained without fundamental alteration to assay protocols. The conjugates are based on novel chemistry that facilitates conjugation of multiple enzyme molecules (alkaline phosphatase or horseradish peroxidase) to a single streptavidin or antibody molecule via a dextran backbone. In most traditional ELISA systems, the conjugates can be substituted directly for the existing HRP or alkaline phosphatase conjugate.
Streptavidin conjugated to multiple HRP molecules on a dextran backbone.

Application

For use in ELISA systems

Features and Benefits

  • It can be used in direct substitution for a standard anti-rabbit HRP conjugate, without the need to alter existing protocols.
  • It is important to optimize the dilution by setting up a range test.
  • Option to reduce incubation times.
  • Well-controlled nonspecific binding properties

Storage and Stability

Please be aware this product may be shipped 30 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

AMDEX is a trademark of Cytiva

Storage Class Code

12 - Non Combustible Liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service