Skip to Content
Merck
All Photos(1)

Key Documents

EHU130891

Sigma-Aldrich

MISSION® esiRNA

targeting human TUG1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCAAGATGCCAGTTTTTGCTTCTTTGTTAGTTGTCAGCTGCTTTTATCAAATTTCAGGCCATTATCCAACAAACACTATAAAAATGTTTGAACAATTGGATTTCAAACATTTTCGTTTTGTGGAGTGGTGCTCACCAAGTGGTACAGCCCTAAGCAAGTGAACACAAACACATTTAAGTGTATTTTGTCTGATTAGATGTTAGCCAGTTATGCTATTTCATTCAAATGTCTGAAAAAATCAATTGACTATTCCCTTTTCCTAAAGGGCAGAGACAGATAATCTCACTTCCAGAGAAATGACTTGGAGAAAAAAAAGTGTTGGTCTTTTTGCTCTTTTGTAATTAAATCCGGATGTACCTCAAAAGACTTAAGACTGTGGTGATAAGATGCTTTCCTCAGCAGAAAGGAGGGAAAAAAAACAACTGGAACTCAAAGCTTGAAATTCTGTGGCAAAACATGAGATGTCCAGGATTGGAGGTTGAA

Ensembl | human accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

human ... TUG1(55000)

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Xu et al.
European review for medical and pharmacological sciences, 23(11), 4698-4705 (2019-06-19)
To explore the correlation between plasma level of lncRNA TUG1 with PSA level, Gleason grading and tumor node metastasis (TNM) stage of prostate cancer (PCa) patients. This study aims to evaluate the potential diagnostic and prognostic values of TUG1 in
Pan Wang et al.
Journal of experimental & clinical cancer research : CR, 39(1), 7-7 (2020-01-11)
Long noncoding RNAs (lncRNAs) are involved in the progression of various cancers and affect the response to radiotherapy. This study focused on clarifying the underlying mechanism by which lncTUG1 affects the radiosensitivity of esophageal squamous cell carcinoma (ESCC). lncTUG1, miR-144-3p
Xiaodi Liu et al.
Journal of cellular biochemistry (2019-03-06)
This study intended to investigate the potential molecular mechanism of long noncoding RNA (lncRNA) taurine upregulated gene 1 (TUG1) in the development of sepsis-associated acute kidney injury (AKI). The expression of TUG1 in the serum from patients with sepsis resulted
Dong Lv et al.
OncoTargets and therapy, 13, 5857-5868 (2020-07-02)
Clear cell renal cell carcinoma (ccRCC) is a common urological carcinoma in adults. Long non-coding RNA (lncRNA) taurine-upregulated gene 1 (TUG1) has been reported to be involved in the progression of diverse human cancers, including renal cell carcinoma (RCC). However
Yanqian Wang et al.
International journal of oncology, 54(4), 1317-1326 (2019-02-06)
Melanoma is an aggressive type of skin cancer, characterized by high mortality rates worldwide. Therefore, the identification of new diagnostic markers and therapeutic targets for melanoma is imperative. Accumulating evidence has demonstrated that long non-coding RNAs (lncRNAs) play important roles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service