CLS352057
Corning® Falcon® Round Bottom Test Tubes
capacity 14 mL, cap: no, sterile, case of 500
Synonym(s):
culture tube
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41121700
NACRES:
NB.13
Recommended Products
material
round polystyrene
sterility
sterile
feature
cap: no
packaging
case of 500
manufacturer/tradename
Corning 352057
capacity
14 mL
Looking for similar products? Visit Product Comparison Guide
Application
Liquid Handling
Legal Information
Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service