Skip to Content
Merck
All Photos(1)

Key Documents

293 N3S

92052131, human kidney, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

293 N3S, 92052131

biological source

human kidney

growth mode

Suspension

karyotype

Not specified

morphology

Not specified

products

Adenovirus 5 early region (E1) antigens

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human embryo kidney

Cell Line Description

Adapted for growth in suspension by passaging 293 cells in nude mice. The cells express adenovirus 5 early region 1 (E1) antigens and are permissive for adenovirus 5 host range mutants defective in E1 functions.

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 11
D13S317: 12,14
D16S539: 9
D5S818: 9
D7S820: 11,12
THO1: 7,9.3
TPOX: 11
vWA: 16,19

Culture Medium

Joklik′s MEM + 2mM Glutamine + 10% Horse Serum.

Subculture Routine

Maintain cultures between 5 x 10,000 to 5 x 100,000 cells/ml in a Techne stirrer; 5% CO2; 37°C. Cell doubling time is 30 hours.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service