Skip to Content
Merck
All Photos(1)

Key Documents

HL60 15-12

88120805, human blood, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

HL60 15-12, 88120805

biological source

human blood

growth mode

Suspension

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian promyelocytic leukaemia

Cell Line Description

HL60 15-12 is a variant of HL60 which is capable of differentiating towards either neutrophils or monocytes with treatment of appropriate agents.

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS) + 1.25% Dimethyl Sulphoxide (DMSO).

Subculture Routine

Maintain cultures between 2x100,000 to 1,000,000 cells/ml. NB: The cells will differentiate if the culture medium is allowed to become acidic. 5% CO2; 37°C. Freeze in 10% glycerol and 90% FBS.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service