Skip to Content
Merck
All Photos(1)

Key Documents

49225

Sigma-Aldrich

α-D-Glucose 1,6-bisphosphate potassium salt hydrate

≥99.0% (TLC)

Synonym(s):

α-D-Glucose 1,6-diphosphate potassium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10K4O12P2 · xH2O
CAS Number:
Molecular Weight:
492.48 (anhydrous basis)
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥99.0% (TLC)

form

solid

impurities

≤15% water

color

white

solubility

H2O: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

[K+].[K+].[K+].[K+].O[C@H]1[C@H](O)[C@@H](COP([O-])([O-])=O)O[C@H](OP([O-])([O-])=O)[C@@H]1O

InChI

1S/C6H14O12P2.4K/c7-3-2(1-16-19(10,11)12)17-6(5(9)4(3)8)18-20(13,14)15;;;;/h2-9H,1H2,(H2,10,11,12)(H2,13,14,15);;;;/q;4*+1/p-4/t2-,3-,4+,5-,6-;;;;/m1..../s1

InChI key

OVRXVCAVDXBPQT-QMKHLHGBSA-J

Looking for similar products? Visit Product Comparison Guide

Application

α-D-Glucose 1,6-bisphosphate is used in the identification, differentiation and characterization of α-phosphoglucomutase(s), β-phosphoglucomutase(s) and α- and β-hexoglucomutase(s).

Biochem/physiol Actions

Metabolite formed via the activity of Glucose-1,6-biphosphate synthase. Glucose-1,6-biphosphate (G-1,6-BP) acts as an inhibitor of hexokinase function and an activator of phosphofructokinase-1 and pyruvate kinase. G-1,6-BP also acts as a coenzyme for phosphoglucomutate and a cofactor for phosphopentomutase.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jessica P Lao et al.
G3 (Bethesda, Md.), 9(2), 413-423 (2018-12-12)
Phosphomannomutase 2 Deficiency (PMM2-CDG) is the most common monogenic congenital disorder of glycosylation (CDG) affecting at least 800 patients globally. PMM2 orthologs are present in model organisms, including the budding yeast Saccharomyces cerevisiae gene SEC53 Here we describe conserved genotype-phenotype

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service