Skip to Content
Merck
All Photos(2)

Key Documents

NIST999C

Potassium chloride

Primary Standard, NIST® SRM®

Synonym(s):

Potassium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KCl
CAS Number:
Molecular Weight:
74.55
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

product name

Potassium Chloride Primary Standard, NIST®SRM®

form

powder

packaging

pkg of 30 g

manufacturer/tradename

NIST®

mp

770 °C (lit.)

density

1.98 g/mL at 25 °C (lit.)

application(s)

general analytical

storage temp.

15-25°C

SMILES string

[Cl-].[K+]

InChI

1S/ClH.K/h1H;/q;+1/p-1

InChI key

WCUXLLCKKVVCTQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is intended for use as an analytical standard of known potassium (K) and chloride (Cl?) content. This lot of potassium chloride (KCl) was prepared to ensure a material of high purity and homogeneity and has been assayed after drying at 500 °C. A unit of SRM 999c consists of a single glass bottle containing 30 g of the material.

SRM 999C_cert

SRM 999C_SDS

Application

May be used for the preparation of phosphate buffered saline, and for the extraction and solubilization of proteins.

Other Notes

Certified for the analytes listed below.
Potassium (K), Chlorine (Cl), Potassium chloride (KCl)

Reference/Informational value is provided for the following analytes.
Bromine (Br), Alkalinity (as KOH), Aluminum (Al), Calcium (Ca), Fluorine (F), Sodium (Na), Rubidium (Rb), Sulfur (S), Silicon (Si)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Explore the tests integral to the instrument qualification process for ion chromatography instruments. Learn about the effectiveness of a pre-packaged IQ/OQ/PQ kit, featuring six certified bromide standard solutions for comprehensive IC instrument qualification tests.

Explore the tests integral to the instrument qualification process for ion chromatography instruments. Learn about the effectiveness of a pre-packaged IQ/OQ/PQ kit, featuring six certified bromide standard solutions for comprehensive IC instrument qualification tests.

Explore the tests integral to the instrument qualification process for ion chromatography instruments. Learn about the effectiveness of a pre-packaged IQ/OQ/PQ kit, featuring six certified bromide standard solutions for comprehensive IC instrument qualification tests.

Explore the tests integral to the instrument qualification process for ion chromatography instruments. Learn about the effectiveness of a pre-packaged IQ/OQ/PQ kit, featuring six certified bromide standard solutions for comprehensive IC instrument qualification tests.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service