Skip to Content
Merck
All Photos(1)

Key Documents

65690

Sigma-Aldrich

3-(Methylamino)propylamine

technical, ≥97.0% (GC)

Synonym(s):

N-Methyl-1,3-propanediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3NH(CH2)3NH2
CAS Number:
Molecular Weight:
88.15
Beilstein:
878143
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Quality Level

Assay

≥97.0% (GC)

refractive index

n20/D 1.447 (lit.)
n20/D 1.447

bp

139-141 °C (lit.)

density

0.844 g/mL at 25 °C (lit.)

SMILES string

CNCCCN

InChI

1S/C4H12N2/c1-6-4-2-3-5/h6H,2-5H2,1H3

InChI key

QHJABUZHRJTCAR-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

107.6 °F - DIN 51755 Part 1

Flash Point(C)

42 °C - DIN 51755 Part 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service