Skip to Content
Merck
All Photos(1)

Key Documents

AGRLE-RO

Roche

Agarose LE

low electroendosmosis

Synonym(s):

agarose

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

biological source

algae (seaweed)

form

powder

packaging

pkg of 100 g (11685660001)
pkg of 500 g (11685678001)

manufacturer/tradename

Roche

EEO

0.05 -0.13

transition temp

gel point 36 °C ±1.5

gel strength

>=2,500 g/cm2

foreign activity

Dnase, none detected
Rnase, none detected

shipped in

ambient

storage temp.

20-25°C

General description

Agarose is obtained from seaweed and contains repeated agarobiose units. During agarose gel formation, the polymers aggregate to form a network with varying pore sizes. This property is used in agarose gel electrophoresis to separate DNA.

Application

Agarose LE has been used for:

  • analyzing fragments between 0.2 and 15 kb
  • analysis of PCR products
  • examination of restriction endonucleases
  • digests of plasmid, cosmid, and λ phage DNA
  • electrophoresis of RNA in, e.g., denaturing gels containing formaldehyde
  • in combination with ethidium bromide ​in DNA electrophoresis to facilitate the separation and visualization of DNA fragments.
Nucleic acid fragments separated with Agarose LE can be blotted to nylon or nitro-cellulose membranes by all standard blotting techniques.
Important Note: Detection with nonradioactive probes e.g., digoxigenin-labeled nucleic acids, does not interfere with the use of Agarose LE.

Quality

Absence of DNase: none detected according to the current Quality Control procedures.
Absence of RNase: none detected according to the current Quality Control procedures.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

"Impact of Interleukin 16 (IL-16) Gene Polymorphism among

Seropositive Stages in HIV-1 Infected Patients in North India.
Kakkar K
Journal of Virology & Antiviral Research, 8, 006-011 (2016)
Emily Hodges et al.
Nature protocols, 4(6), 960-974 (2009-05-30)
Complementary techniques that deepen information content and minimize reagent costs are required to realize the full potential of massively parallel sequencing. Here, we describe a resequencing approach that directs focus to genomic regions of high interest by combining hybridization-based purification
Pei Yun Lee et al.
Journal of visualized experiments : JoVE, 62(62), 3923-3923 (2012-05-02)
Agarose gel electrophoresis is the most effective way of separating DNA fragments of varying sizes ranging from 100 bp to 25 kb(1). Agarose is isolated from the seaweed genera Gelidium and Gracilaria, and consists of repeated agarobiose (L- and D-galactose)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service