Key Documents
ZLXL00P20
Milli-Q
Loop-20 m tubing 10x16 with Inters
For addition of single remote Point Of Delivery (POD) on the water distribution loop
About This Item
UNSPSC Code:
41104214
NACRES:
JA.13
packaging
pkg of 1 unit
L
20 m , Tube
compatibility
for use with Milli-Q® CLX
for use with Milli-Q® HR
for use with Milli-Q® HX
General description
This 20 m loop option is used to provide a single remote Point Of Delivery (POD) on the water distribution loop.
This kit contains 20 meters of 10x16 mm tubing plus parts needed to make one Point Of Delivery. The Point Of Delivery could be used to feed water to an analyzer or other device.
Legal Information
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany