Skip to Content
Merck
All Photos(1)

Documents

MABF848

Sigma-Aldrich

Anti-EBI3 Antibody/IL-35, clone V1.4C4.22 (Neutralizing)

clone V1.4C4.22, from mouse

Synonym(s):

EBI3/IL-35, IL-27B, Interleukin-27 subunit beta, IL-27 subunit beta, IL-27B, Epstein-Barr virus-induced gene 3 protein homolog

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

V1.4C4.22, monoclonal

species reactivity

mouse

technique(s)

neutralization: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Ebi3(50498)

General description

EBI3/IL27B is the beta subunit of the functional cytokine IL-27. The beta subunit is required for activity. IL-27 is a critical cytokine of mammalian innate immune response. IL-27 has both pro and anti-inflammatory activities. IL-27 regulates T-helper cell development and differentiation into TH1, TH2 and TH17 effector cells. IL-27 also is important in isotype switching for B-cells, and it drives rapid clonal expansion of native but not memory CD4 cells. IL-27 is also important for its anti-tumor and anti-angiogenic activity on tumor and cancer cells. The beta subunit EBI3/IL27B is also a component of IL-35 which is a heterodimer of IL-12alpha and IL-27B/EBI3. IL-35 plays a role in immune suppression. IL-27 is a secreted cytokine, thus IL-27beta subunit is expressed intracellularly associated with the endoplasmic reticulum and membrane vesicles and is detected extracellularly as part of the mature cytokine as well.

Specificity

Clone V1.4H6.25 (MABF836) was used in in-vitro mIL-35 neutralizing studies and clone V1.4C4.22 (MABF848) was used in in-vivo mIL-35 neutralizing studies (Collison, L.W., et al. (2010). Nat Immunol. 11(12):1093-1101).

Immunogen

Recombinant protein corresponding to mouse EBI3/IL-35.

Application

Anti-EBI3 Antibody/IL-35, clone V1.4C4.22 (Neutralizing) is an antibody against EBI3 /IL-35 for use in Western Blotting, Neutralizing.
Neutralizing Analysis: A representative lot detected EBI3/IL-35 during in-vitro neutralizing (Collison, L.W., et al. (2010). Nat Immunol. 11(12):1093-1101).
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

Evaluated by Western Blotting in recombinant m-EBI3/IL-27.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected EBI3/IL-35 in 1 µg of recombinant m-EBI3/IL-27.

Target description

~25 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naifu Wan et al.
Annals of translational medicine, 9(11), 926-926 (2021-08-06)
To explore the source, the role and the specific mechanism of IL-35 and its downstream molecules in the development of pulmonary hypertension. 8-10 weeks male mice were undergoing hypoxia combined with SU5416 (HySu) to establish a pulmonary hypertension (PH) model.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service