IBFP0813C
Immobilon® Blotting Filter Paper
100 sheets, 8.5 cm x 13.5 cm, Chromotography-Grade Blotting Paper for blotting
Synonym(s):
Western blot filter paper, blotting paper
About This Item
Recommended Products
product name
Immobilon® Blotting Filter Paper, 8.5 x 13.5 cm sheet, Chromotography-Grade Blotting Paper for western blotting.
material
plain filter
white filter
Quality Level
feature
hydrophilic
manufacturer/tradename
Immobilon®
technique(s)
dot blot: suitable
western blot: suitable
filter L × W
8.5 cm × 13.5 cm
thickness
550 μm
compatibility
for use with Amido black
for use with CPTS
for use with Colloidal gold
for use with India ink
for use with Ponceau-S red
detection method
chemiluminescent
colorimetric
fluorometric
shipped in
ambient
General description
Application
Legal Information
Storage Class Code
10-13 - German Storage Class 10 to 13
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service