Skip to Content
Merck
All Photos(2)

Key Documents

203646

Sigma-Aldrich

DMH1

≥97% (HPLC), solid, BMP inhibitor, Calbiochem®

Synonym(s):

BMP Inhibitor II, DMH1, ALK2/3 Inhibitor, DMH1, Bone Morphogenetic Protein Inhibitor, DMH1, 4-(6-(4-Isopropoxyphenyl)pyrazolo[1,5-a]pyrimidin-3-yl)quinoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H20N4O
CAS Number:
Molecular Weight:
380.44
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

product name

BMP Inhibitor II, DMH1, The BMP Inhibitor II, DMH1, also referenced under CAS 1206711-16-1, controls the biological activity of BMP. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange

solubility

DMSO: 16.6 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C24H20N4O/c1-16(2)29-19-9-7-17(8-10-19)18-13-26-24-22(14-27-28(24)15-18)20-11-12-25-23-6-4-3-5-21(20)23/h3-16H,1-2H3

InChI key

JMIFGARJSWXZSH-UHFFFAOYSA-N

General description

A cell-permeable, potent, and highly selective BMPR (bone morphogenetic protein receptor) inhibitior (IC50 against human ALK2/BMPR-I = 107.9 nM) that exhibits no inhibitory activity against ALK5, AMPK, FLK1/KDR/VEGRF2, or PDGFRβ. Effectively blocks ALK2- and ALK3-, but not ALK6-, mediated transcription activity (IC50<500 nM, <100 nM, and >10 M in reporter assays using C2C12BRA expressing constitutively active ALK2, ALK3, or ALK6, respectively). Comparing to its non-BMPR-selective structural analog dorsomorphin, DMH1 is 12.5-times more potent in preventing BMP pathway-dependent DV (dorsoventral) development (EC100 = 0.2 M DMH1 or 2.5 M DM) in zebrafish embryo in vivo, while being non-toxic to the fish embryo and exhibiting no effect against VEGF signaling-dependent intersomitic vessel (ISV) formation even at concentrations as high as 50 M.
A cell-permeable, potent, and highly selective BMPR inhibitior (IC50 against human ALK2/BMPR-I = 107.9 nM), exhibiting no activity toward ALK5, ALK6, AMPK, FLK1/KDR/VEGRF2, or PDGFRβ. Comparing to its non-BMPR-selective structural analog dorsomorphin DMH1 is 12.5-times more potent in preventing BMP pathway-dependent dorsoventral development (EC100 = 0.2 M) in zebrafish embryo in vivo, while being non-toxic to the fish embryo and exhibiting no effect against VEGF signaling-dependent intersomitic vessel formation even at concentrations as high as 50 M.

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Hao, J., et al. 2010. ACS Chem.Biol.5, 245.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eduardo Branco de Sousa et al.
Revista brasileira de ortopedia, 57(2), 314-320 (2021-03-31)
Objective  Our goal was to evaluate the modulation of the synovial fluid cells (SFC) from patients with and without osteoarthritis (OA) by bone morphogenetic protein 4 (BMP-4), Smad-3 and transforming growth factor beta (TGF-β). Methods  Synovial fluid was collected from
Transient suppression of SUMOylation in embryonic stem cells generates embryo-like structures.
Cossec, et al.
Cell Reports, 42, 112380-112380 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service