Skip to Content
Merck
All Photos(2)

Key Documents

860735E

Avanti

3-O-sulfo-galactosyl(β) sphingosine-d7 (d18:1) ammonium salt

Avanti Research - A Croda Brand

Synonym(s):

3-O-sulfo-D-galactosyl(β) D-erythro-sphingosine-d7 (d18:1, ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H43D7N2O10S
Molecular Weight:
565.77
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

ethanol solution

packaging

package of 1 × 100 μg ((860735E-100UG))

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCC([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])(O)[C@@]([H])(N)CO[C@H](O1)[C@H](O)[C@@H](OS([O-])(=O)=O)[C@H]([C@H]1CO)O.[NH4+]

Packaging

2 mL Amber Glass Sealed Ampule (860735E-100UG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service