Key Documents
SYNF122450
Synthware single-neck flat-bottom flask
capacity 50 mL, joint: ST/NS 24/40
Synonym(s):
Synthware F122450, glass round flask
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
material
borosilicate glass flask
flat bottom
single-neck
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F122450
capacity
50 mL
General description
Heavy wall single neck, flat bottom flask with an outer joint. Heavy wall design. All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
Legal Information
Synthware is a trademark of Kemtech America Inc.