Skip to Content
Merck
All Photos(1)

Key Documents

BR23766

BRAND® SILBERBRAND automatic burette, Dr. Schilling

burette size 25 mL, bottle size 1000 mL, amber AR-Glass

Synonym(s):

Automatic Burettes, Burettes, Dr. Schilling pattern, SILBERBRAND

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121808
NACRES:
NB.43

material

amber AR-Glass

feature

Schellbach stripe
graduated in white

manufacturer/tradename

BRAND 23766

volume measuring range

25 mL, accuracy: ±0.08 mL

bottle size

1000 mL

burette size

25 mL

Looking for similar products? Visit Product Comparison Guide

General description

SILBERBRAND Schellbach Stripe - Short marks at major graduations. Reading at point of contact of the two tips.

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service