Merck
HomeChemical Analysis for Food and Beverage TestingDetermination of Urea in Milk by HPTLC
Determination of Urea in Milk by HPTLC
Sanjay Poman, Application Scientist
Abstract
This article details a quick HPTLC method to detect urea in milk using a TLC plate scanner and a Silica Gel 60 F254 HPTLC Plate. The results indicate that none of the four milk samples tested exceeded the acceptable limit of 70 mg/100 mL, demonstrating the method’s efficacy for quick and reliable detection of urea, thus offering a robust tool for quality control and food safety in dairy products.
Section Overview
Introduction
Urea also known as carbamide is an organic compound serves an important role in the metabolism of nitrogen containing compounds by animals and is the main nitrogen containing substance in the urine of mammals. Urea is widely used in fertilizers as a source of Nitrogen and is important raw material for the chemical industry. Milk is an important food component in our regular diet. Urea in cow‘s milk varies from 20 mg to 70 mg/100 mL of urea and urea content above this range is said to be adulterated or deliberately added.1 These unhygienic materials are mainly added to compensate the expenses and hence to yield higher profit by cheap and low quality adulterants in order to increase the solid non-fat value in milk.
The present study illustrates a new method based on thin layer chromatography (TLC) using HPTLC plates to establish a quick limit test for the detection of urea in milk.
Experimental
The four locally sourced milk samples were investigated. Each sample (5 mL) was diluted with 5 mL of water and then centrifuged at 2500 rpm for 10 minutes. The supernatants were then used for analysis. The standard with 0.7 mg/mL urea (representing the limit concentration of 70 mg/100 mL) was prepared in water/methanol (1:9) v/v. The analysis was performed under conditions described in Table 1.
Standards & Sample Solution
Sample preparation:
Dissolve 5 mL of milk in 5 mL of water. Centrifuge at 2500 rpm for 10 minutes and take supernatant for analysis.
Standard preparation:
Weigh urea 0.7 mg/mL in a suitable volumetric flask using (1:9 ) v/v water / methanol as diluent
Chromatographic Conditions
Plate:
HPTLC plate Silica Gel 60 F254, 20 x 10 cm plates (1.05642)
Sample application:
Band wise with ATS4 from Camag: 2.5 µL of the Reference solution three times, 5.0 µL of the test solutions A to D three times
Mobile phase:
1-propanol/water (8:2) v/v
Migration distance:
8 cm
Drying:
At room temperature
Derivatization:
Spray with Ehrlich reagent (1- (Dimethyl amino)- Benzaldehyde Hydrochloride (Dissolve 500 mg of 1-(Dimethyl amino) Benzaldehyde in 40 ml methanol and treat it with cooling 10 ml HCl (32%). The temperature should not fall below 20 °C or rise above 40 ºC.
Detection:
Scanning at 440 nm with TLC Scanner 4 from Camag
Table 1.TLC conditions for determination of urea in milk
Results
The developed plate is shown in Figure 1. The principal spot in the chromatogram obtained with the test solution is similar in position, color & size to the principal spot in the chromatogram obtained with the reference solution.
A derivatized HPTLC plate, viewed under white light, appears in a pale yellow hue. On both edges of the plate, there are 9 black markings, starting from the bottom line used for spotting, labeled from 0.0 to 0.9 at 0.1 cm intervals. Along the bottom, labels 1 to 10 are shown from left to right, with labels 1 to 4 in black and 5 to 10 in pink, representing the ten spots applied. The plate displays ten bright yellow-colored spots between Rf values of 0.45 to 0.55, corresponding to tracks labeled 1 to 10 from left to right. The first three spots on the left are for urea reference solution at a concentration of 0.7 mg/mL, followed by four sets of three spots each for four different milk samples analyzed.
Figure 1.Derivatized HPTLC plate under in white light illumination (pink marked milk sample tracks quantified by densitometry see Table 2)
Examples of spot scans for the reference solution and a milk sample are shown in Figure 2 & 3. Retardation factor for reference solution and Test solution is 0.50 & 0.48 respectively indicating similar position and color.
A TLC-densitogram obtained using a TLC analyzer for urea reference solution, shows the retention factor on the x-axis with major tick marks at intervals of 0.1, ranging from 0.0 to 1.0. The y-axis, representing detector response in absorbance units (AU), has 5 major tick marks from 0.10 to 0.50, at intervals of 0.10. The densitogram displays one blue-colored peak labeled 1, which begins to form at an Rf value of 0.4, reaches its maximum at 0.50, and then declines, disappearing at 0.55.
Figure 2.Chromatographic Data Example - Reference Solution
Chromatographic Result Example - Reference Solution
        Compound
Rf
Area
1
         Urea
0.50
2640
A TLC-densitogram obtained using a TLC analyzer for urea test solution, shows the retention factor on the x-axis with major tick marks at intervals of 0.1, ranging from 0.0 to 1.0. The y-axis, representing detector response in absorbance units (AU), has 7 major tick marks from 0.05 to 0.35, at intervals of 0.05. The densitogram displays one blue-colored peak labeled 1, which begins to form at an Rf value of 0.4, reaches its maximum at 0.48, and then declines, disappearing at 0.55.
Figure 3.Chromatographic Data Example - Test Solution
Chromatographic Result Example - Test Solution
        Compound
Rf
Area
1
         Urea
0.48
1836
As examples 2 spots of each milk sample were quantified (Figure 4 & Table 2). The 4 different commercial milk samples tested as per the illustrated method showed urea concentration less than 1.75 µg on the plate or less than 0.7 mg/mL sample (700 ppm).
A calibration curve plot for urea and samples, scanned at 440 nm, shows detector response (AU) versus quantity (µg). The x-axis has eight major ticks from 0.25 to 2.00 at intervals of 0.25, representing quantity in µg. The y-axis, showing detector response in AU, has six major ticks from 0.005 to 0.03 at intervals of 0.005. A straight grey line originates from the origin, turning red within the calibration range. Sample values are represented by sky blue points, while reference values are marked with mint green X-marks on the line.
Figure 4.Applied calibration for urea and samples (scan at 440 nm). Reference point (reference solution) of 1.75 µg on plate is equivalent to 70 mg/100mL (700 ppm) in milk.
Track
Sample
Rf
Amount on Plate (µg)
Concentration in Milk Sample (mg/100 mL)
5
A
0.482
1.151
46.04
6
A
0.485
1.220
48.80
7
B
0.492
1.249
49.96
8
B
0.495
1.235
49.40
10
C
0.487
0.8441
33.76
12
C
0.488
0.8597
34.39
14
D
0.493
1.166
46.64
15
D
0.504
1.166
46.64
Table 2.Determined the concentrations of 2 of the 3 spots of each sample is shown as an example
Conclusion
The here displayed method was used for a quick limit check of urea content in milk samples by HPTLC using a plate scanner for quantification. Four samples could be investigated on one plate, and none exceeded the limit value of 70 mg/100 mL. The HPTLC provides therefore a simple and quick option for the determination of urea content and possible food fraud in multiple milk samples in parallel.
Find more applications on Food & Beverage Testing.
Related Products
Si è verificato un errore inatteso.
Network error: Failed to fetch
References
1.
Azad T, Ahmed S. 2016. Common milk adulteration and their detection techniques. FoodContamination. 3(1): https://doi.org/10.1186/s40550-016-0045-3