Merck
HomeSmall Molecule HPLCTanaka Test for C18 Stationary Phase in HPLC Columns
Tanaka Test for the Characterization of C18 Stationary Phase in HPLC Columns
Introduction
The properties of stationary phases, both physical and chemical, play a crucial role in the performance of HPLC. However, achieving reproducibility requires comprehensive chromatographic characterization. Consequently, prominent HPLC scientists have employed a range of approaches to assess selectivity. The Tanaka Test is widely recognized as the industry standard for evaluating column selectivity and performance.
The test facilitates the comparison of HPLC columns and assists in choosing the most suitable column for specific separations by assessing essential parameters. Seven substances are utilized to assess capacity, hydrophobicity, steric selectivity, and silanophilic characteristics. The parameters are displayed on a hexagonal grid, offering a visual depiction of the sorbent’s characteristics. A hexagon that exhibits greater symmetry and a larger area indicates a more balanced stationary phase in terms of its chromatographic properties.
Parameter/Property
Property of
stationary phase
Factors in preparation of
the stationary phase
A
k (Pentylbenzene)
80% Methanol
Retention
capacity
Amount of alkyl chains
Silica surface
Silica coverage
B
k (Pentylbenzene) /
k (Butylbenzene)
80% Methanol
Hydrophobicity
Hydrophobic capacity
(CH2-group selectivity)
Surface coverage
C
k (Triphenylene) /
k (o-Terphenyl)
80% Methanol
Steric selectivity
Steric (shape) selectivity
Silane functionality
Surface coverage
D
k (Caffeine) / k (Phenol)
30% Methanol
Silanol capacity
Silanol capacity (content and type of silanols)
Residual silanols
Endcapping
Surface coverage
E
k (Benzylamine) / k (Phenol)
30% Methanol /
70% Phosphate buffer pH 7.6
Ion exchange capacity
Ion exchange capacity at
pH 7
Residual silanols
Active sites at pH 7 
F
k (Benzylamine) / k (Phenol)
30% Methanol / 70% Phosphate buffer pH 2.7
Ion exchange capacity
Ion exchange capacity at
pH 3
Active sites at pH 3
Treatment of basic silica (metal impurities
Table 1.Tanaka test parameters for the characterization of revered-phase HPLC columns
Tanaka Tests for Purospher® STAR RP-18 endcapped
Tanaka Test 1: Retention Capacity, Hydrophobicity, Steric Selectivity
The Tanaka 1 Test describes the retention capacity, the hydrophobicity, and the steric selectivity of RP-phases.
HPLC Parameters
Column
Purospher® STAR RP-18 endcapped (5μm) LiChroCART® 150-4,6 HPLC column
Mobile Phase
Methanol / Water 80:20
Flow rate
1.0 mL/min
Detection
UV 254 nm
Temperature
30° C
Inj. Volume
10 μl
Sample
1. Uracil
2. Butylbenzene
3. o-Terphenyl
4. Pentylbenzene
5. Triphenylene
Table 2.HPLC conditions for the Tanaka test 1
Tanaka Test 2: Silanol Capacity
The Tanaka 2 Test illustrates the silanophilic properties of stationary phases. The Phenol/Pyridine test measures silanol activities.
HPLC Parameters
Column
Purospher® STAR RP-18 endcapped (5μm) LiChroCART® 125-4 HPLC column
Mobile Phase
A: Methanol / Water 30:70 (v/v)
B: Acetonitrile / Water 30:70 (v/v)
Flow rate
1.0 mL/min
Detection
UV 254 nm
Sample
A: Uracil (1), Caffeine (2), Phenol (3)
B: Pyridine (1), Phenol (2)
Table 3.HPLC conditions for the Tanaka test 2
Tanaka Test 3 and 4: Ion Exchange Capacities
The results obtained from the Tanaka 3 Test (at low pH) demonstrate whether the deactivation of the residual silanol groups is complete. The Tanaka 4 Test (at high pH) indicates the complete absence of metal in the base sorbent.
HPLC Parameters
Column
Purospher® STAR RP-18 endcapped (5μm) LiChroCART® 125-4 HPLC column
Mobile Phase
Methanol / 0.02 mol/L
Phosphoric acid 30:70 (v/v)
Flow rate
0.6 ml/min
Detection
UV 254 nm
Sample
1. Uracil
2. Benzylamine
3. Phenol
Table 4.HPLC conditions for the Tanaka test 3 and 4
Results
In order to facilitate a clear comparison of the chromatographic properties of various phases, a hexagonal net diagram was implemented to visually depict the measured Tanaka parameters. To standardize the display, the measured values are multiplied by certain factors for this diagram. Table 5 contains the measured, un-modified values and the Figure 1 shows the Tanaka plots for the tested stationary phases, demonstrating their relative performance across the selected parameters.
The Purospher® STAR RP-18 endcapped column demonstrated superior selectivity across all critical parameters in a comparative analysis with other "high-purity" RP-18 endcapped columns, as illustrated in Figure 1. The Tanaka Test results showed that this column offers a balanced performance in terms of hydrophobicity, retention, steric selectivity, and silanol activity, making it a column of choice for a wide range of HPLC separations.
A radar chart (spider chart) with six axes extending outward from a central point. Each axis represents a different chromatographic property or performance metric (such as hydrophobicity, steric selectivity, hydrogen bonding capacity, etc.). Multiple overlapping polygons are drawn on the chart, each one representing the performance of a different HPLC column. The Purospher® STAR RP-18 endcapped column is compared to seven competitor columns, with each column's performance plotted along the axes. The resulting shapes allow for easy visual comparison of how each column performs across the six chromatographic parameters.
Figure 1.Six axes radar Tanaka plots for Purospher® STAR RP-18 endcapped column and seven competitor columns.
A.
K (Pentylbenzene)
9.59
B.
α (Pentyl-/ Butylbenzene)
1.51
C.
α (Triphenylene/ o-Terphenyl)
1.63
D.
α (Caffeine/ Phenol)
0.44
E.
α (Benzylamine/ Phenol; pH 7.6)
0.23
F.
α (Benzylamine/ Phenol; pH 2.7)
0.02
Table 5.Measured Tanaka values without multipliers for Purospher® STAR RP-18 endcapped column
Conclusion
The Tanaka Test is an indispensable tool for chromatographic characterization, as it enables a comprehensive assessment of HPLC columns. The Purospher® STAR RP-18 endcapped column is a robust option for high-performance, reliable separations, with its balanced chromatographic properties confirmed through rigorous testing. By understanding the significance of each parameter assessed in the Tanaka Test, it is possible to make well-informed decisions regarding column selection for particular analytical requirements.
Related Products
Si è verificato un errore inatteso.
Network error: Failed to fetch