Passa al contenuto
MilliporeSigma
GC Analysis of Phenols in Water on a 5% Phenyl Column after SPE using Supelclean ENVI-Chrom P
GC Analysis of Phenols in Water on a 5% Phenyl Column after SPE using Supelclean™ ENVI-Chrom P suitable for GC, application for SPE
Materiali
Colonnina o piastra per SPE
N° Catalogo
Descrizione
Determinazione del prezzo
Colonnine SupelcleanENVI-Chrom P per SPE
bed wt. 250 mg, volume 6 mL, pk of 30
CONDITIONS
sample preparation
SPE (Solid Phase Extraction)
sample/matrix
water spiked with phenol compounds
SPE tube/cartridge
Supelclean ENVI-Chrom P, 250 mg/6 mL (57225-U)
condition
6 mL methyl t-butyl or ethyl acetate; 6 mL acetonitrile. Equilibrate with 6 mL DI water
sample addition
100 mL water sample.
drying
10 min using vacuum
elution
5 mL methyl t-butyl ether acetate dropwise allowing initial 2 mL to soak in SPE bed
column
5% Phenyl Polysiloxane, 15 m × 0.53 mm I.D., 0.50 μm (Supelco equivalent, Equity-5, 28252-U)
oven
65 °C, 10 °C/min to 185 °C (1 min), 20 °C/min to 275 °C (5 min)
detector
FID, 300 °C
carrier gas
helium
injection
1 μL, splitless (45 sec hold)
Descrizione
Note legali
Supelclean is a trademark of Sigma-Aldrich Co. LLC