Merck
  • Dihydrotestosterone levels at birth associate positively with higher proportions of circulating immature/naïve CD5
Dihydrotestosterone levels at birth associate positively with higher proportions of circulating immature/naïve CD5
Scientific reports (2017-11-16)
Anna-Carin Lundell, Inger Nordström, Kerstin Andersson, Anna Strömbeck, Claes Ohlsson, Åsa Tivesten, Anna Rudin
ABSTRACT
Boys present with higher proportions of immature/naïve CD5
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
Mouse IgA ELISA Kit