Merck
A Cr(VI) selective probe based on a quinoline-amide calix[4]arene.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2017-08-12)
Juliane F Ferreira, Izilda A Bagatin
ABSTRACT
A new quinoline-amide calix[4]arene 3-receptor for detection of hazardous anions and cations have been synthesized. The 3-receptor was examined for its sensing properties towards several different anions (Cr
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
Calcium hydride, purum p.a., ≥97.0% (gas-volumetric), powder