Merck
Studies on coumestrol/beta-cyclodextrin association: Inclusion complex characterization.
International journal of pharmaceutics (2008-11-26)
Camila Franco, Liege Schwingel, Ivana Lula, Rubén D Sinisterra, Letícia Scherer Koester, Valquiria Linck Bassani
ABSTRACT
Coumestrol is an estrogenic and antioxidant agent, characterized by its low solubility in aqueous and lipophilic media, once in the aglicone form. In order to improve its solubility in water, coumestrol was associated with beta-cyclodextrin in aqueous media followed by freeze-drying and characterized by SEM, (1)H NMR and molecular modeling. The analysis proved the existence of an inclusion complex, with higher probability of inclusion of the coumestrol B-ring into the wider rim of the beta-cyclodextrin molecule.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
Coumestrol, ≥95.0% (HPLC)