Merck
  • Green micellar HPLC analysis of three angiotensin-converting enzyme inhibitors in their mixtures with hydrochlorothiazide and modeling of their retention behavior by fitting to Foley's model.
Green micellar HPLC analysis of three angiotensin-converting enzyme inhibitors in their mixtures with hydrochlorothiazide and modeling of their retention behavior by fitting to Foley's model.
Journal of separation science (2017-07-20)
Manal Eid, Yasser El-Shabrawy, Rania El-Shaheny
ABSTRACT
We present an environmentally friendly method for the analysis of three angiotensin-converting enzyme inhibitors and hydrochlorothiazide simultaneously using a green micellar eluent for the first time. The chromatographic separation of enalapril maleate, lisinopril dihydrate, benazepril hydrochloride, and hydrochlorothiazide was implemented on an octadecyl silica column with a solution containing sodium dodecyl sulfate (0.12 M), 1-propyl alcohol (10% v/v), triethylamine (0.3% v/v), and H
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Lisinopril dihydrate, European Pharmacopoeia (EP) Reference Standard