- A flexible transparent colorimetric wrist strap sensor.
A flexible transparent colorimetric wrist strap sensor.
Nanoscale (2016-12-21)
Ting Wang, Yunlong Guo, Pengbo Wan, Xiaoming Sun, Han Zhang, Zhongzhen Yu, Xiaodong Chen
PMID27995251
ABSTRACT
A flexible, transparent, and portable wrist strap sensor device has been well developed from a hierarchical polydiacetylene/MoS
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto