Merck
  • Eco-friendly LC-MS/MS method for analysis of multi-class micropollutants in tap, fountain, and well water from northern Portugal.
Eco-friendly LC-MS/MS method for analysis of multi-class micropollutants in tap, fountain, and well water from northern Portugal.
Analytical and bioanalytical chemistry (2016-10-14)
Marta O Barbosa, Ana R Ribeiro, Manuel F R Pereira, Adrián M T Silva
ABSTRACT
Organic micropollutants present in drinking water (DW) may cause adverse effects for public health, and so reliable analytical methods are required to detect these pollutants at trace levels in DW. This work describes the first green analytical methodology for multi-class determination of 21 pollutants in DW: seven pesticides, an industrial compound, 12 pharmaceuticals, and a metabolite (some included in Directive 2013/39/EU or Decision 2015/495/EU). A solid-phase extraction procedure followed by ultra-high-performance liquid chromatography coupled to tandem mass spectrometry (offline SPE-UHPLC-MS/MS) method was optimized using eco-friendly solvents, achieving detection limits below 0.20 ng L-1. The validated analytical method was successfully applied to DW samples from different sources (tap, fountain, and well waters) from different locations in the north of Portugal, as well as before and after bench-scale UV and ozonation experiments in spiked tap water samples. Thirteen compounds were detected, many of them not regulated yet, in the following order of frequency: diclofenac > norfluoxetine > atrazine > simazine > warfarin > metoprolol > alachlor > chlorfenvinphos > trimethoprim > clarithromycin ≈ carbamazepine ≈ PFOS > citalopram. Hazard quotients were also estimated for the quantified substances and suggested no adverse effects to humans. Graphical Abstract Occurrence and removal of multi-class micropollutants in drinking water, analyzed by an eco-friendly LC-MS/MS method.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Supelco
Chlorfenvinphos, PESTANAL®, analytical standard