Merck
  • Inhibition of macrophage migration inhibitory factor (MIF) tautomerase activity suppresses microglia-mediated inflammatory responses.
Inhibition of macrophage migration inhibitory factor (MIF) tautomerase activity suppresses microglia-mediated inflammatory responses.
Clinical and experimental pharmacology & physiology (2016-10-21)
Yu Zhang, Ruinan Gu, Jia Jia, Tingjun Hou, Long Tai Zheng, Xuechu Zhen
ABSTRACT
Macrophage migration inhibitory factor (MIF), a pleiotropic pro-inflammatory cytokine, is a key regulator in both innate and acquired immunity systems. MIF has become a promising drug target for inflammatory diseases. Apart from its cytokine activities, MIF is known to act as a d-dopachrome tautomerase. Our previous work has identified that 3-[(biphenyl-4-ylcarbonyl)carbamothioyl]amino benzoic acid (Z-590) exhibited a potent inhibitory activity against MIF. In this study, we investigate the effect of Z-590 on lipopolysaccharide (LPS)-activated microglial cell activation. Our results demonstrate that Z-590 significantly decreases the production of nitric oxide (NO), tumour necrosis factor-alpha (TNF-α), interleukin (IL)-6, IL-1β, cyclooxygenase (COX-2), inducible nitric oxide synthase (iNOS) as well as reactive oxygen species (ROS) involved in inhibiting MAKPs signalling pathway in LPS-stimulated microglia cells. Furthermore, Z-590 reduced cytotoxicity of activated microglia toward HT-22 hippocampal cells in a microglia-conditioned medium system. Taken together, these results indicate that MIF inhibitor Z-590 elicits a potent inhibitor for microglia-mediated neuroinflammation.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
MISSION® esiRNA, targeting human MIF