- Determination of rifampicin in human plasma by high-performance liquid chromatography coupled with ultraviolet detection after automatized solid-liquid extraction.
Determination of rifampicin in human plasma by high-performance liquid chromatography coupled with ultraviolet detection after automatized solid-liquid extraction.
Biomedical chromatography : BMC (2016-06-10)
B Louveau, C Fernandez, N Zahr, H Sauvageon-Martre, P Maslanka, P Faure, S Mourah, L Goldwirt
PMID27280327
ABSTRACT
A precise and accurate high-performance liquid chromatography (HPLC) quantification method of rifampicin in human plasma was developed and validated using ultraviolet detection after an automatized solid-phase extraction. The method was validated with respect to selectivity, extraction recovery, linearity, intra- and inter-day precision, accuracy, lower limit of quantification and stability. Chromatographic separation was performed on a Chromolith RP
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto