Merck
Linoleic acid derivative DCP-LA improves learning impairment in SAMP8.
Neuroreport (2005-12-20)
Takahiro Yaguchi, Tetsu Nagata, Takeshi Mukasa, Hirokazu Fujikawa, Hideyuki Yamamoto, Satoshi Yamamoto, Hiroyuki Iso, Akito Tanaka, Tomoyuki Nishizaki
ABSTRACT
In the water-maze test, the linoleic acid derivative, 8-[2-(2-pentyl-cyclopropylmethyl)-cyclopropyl]-octanoic acid (DCP-LA) (1 mg/kg, intraperitoneally), significantly shortened the prolonged latency for accelerated-senescence-prone mice 8 (SAMP8), reaching a level similar to the latency for accelerated-senescence-resistant mice 1 (SAMR1) as control. In the open-field test to assess motor activity, it was confirmed that the DCP-LA effect is not due to increased motor activity. In the passive avoidance test to assess fear memory, DCP-LA had no effect on the latency of acquisition and retention for SAMP8. The results of the present study, thus, suggest that DCP-LA could improve age-related learning impairment by enhancing cognitive functions.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
DCP-LA, ≥95% (HPLC), oil