Merck
  • Optimized alkylated cyclodextrin polysulphates with reduced risks on thromboembolic accidents improve osteoarthritic chondrocyte metabolism.
Optimized alkylated cyclodextrin polysulphates with reduced risks on thromboembolic accidents improve osteoarthritic chondrocyte metabolism.
Rheumatology (Oxford, England) (2011-02-25)
Sara Groeneboer, Stijn Lambrecht, Aad Dhollander, Peggy Jacques, Bert Vander Cruyssen, Rik J Lories, Katrien Devreese, Koen Chiers, Dirk Elewaut, Gust Verbruggen
ABSTRACT
To compare the ability of different cyclodextrin polysulphate (CDPS) derivatives to affect human articular cartilage cell metabolism in vitro. OA chondrocytes were cultured in alginate and exposed to 5 µg/ml of 2,3,6-tri-O-methyl-β-cyclodextrin (ME-CD), 2,3-di-O-methyl-6-sulphate-β-cyclodextrin (ME-CD-6-S), 2,6-di-O-methyl-3-sulphate-β-cyclodextrin (ME-CD-3-S), (2-carboxyethyl)-β-CDPS (CE-CDPS), (2-hydroxypropyl)-β-CDPS (HP-CDPS), 6-monoamino-6-monodeoxy-β-CDPS (MA-CDPS) or β-CDPS for 5 days. Effects on IL-1-driven chondrocyte extracellular matrix (ECM) metabolism were assayed by analysis of the accumulation of aggrecan in the interterritorial matrix, IL-6 secretion and qPCR. MA-CDPS, HP-CDPS, CE-CDPS and CDPS were analysed for their in vitro effect on coagulation and their ability to activate platelets in an in vitro assay to detect possible cross-reactivity with heparin-induced thrombocytopenia (HIT) antibodies. The monosulphated cyclodextrins ME-CD-6-S and -3-S failed to affect aggrecan synthesis and IL-6 secretion by the OA chondrocytes. Polysulphated cyclodextrins MA-CDPS, HP-CDPS, CE-CDPS and CDPS at 5 µg/ml concentrations, on the other hand, significantly induced aggrecan production and repressed IL-6 release by the chondrocytes in culture. aPTT and PT for all derivatives were lengthened for polysaccharide concentrations >50 µg/ml. Five micrograms per millilitre of β-CDPS concentrations that significantly modulated ECM ground substance production in vitro did not affect aPTT or PT. Furthermore, CE-CDPS, in contrast to MA-CDPS, HP-CDPS and CDPS, did not significantly activate platelets, suggesting a minimal potential to induce HIT thromboembolic accidents in vivo. CE-CDPS is a new, structurally adjusted, sulphated β-cyclodextrin derivative with preserved chondroprotective capacity and a promising safety profile.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
D-Glucose-12C6, 99.5 atom % 12C