Merck
  • Ewing sarcoma protein ewsr1 maintains mitotic integrity and proneural cell survival in the zebrafish embryo.
Ewing sarcoma protein ewsr1 maintains mitotic integrity and proneural cell survival in the zebrafish embryo.
PloS one (2007-10-04)
Mizuki Azuma, Lisa J Embree, Hatem Sabaawy, Dennis D Hickstein
ABSTRACT
The Ewing sarcoma breakpoint region 1 gene (EWSR1), also known as EWS, is fused to a number of different partner genes as a result of chromosomal translocation in diverse sarcomas. Despite the involvement of EWSR1 in these diverse sarcomas, the in vivo function of wild type EWSR1 remains unclear. We identified two zebrafish EWSR1 orthologues, ewsr1a and ewsr1b, and demonstrate that both genes are expressed maternally, and are expressed ubiquitously throughout zebrafish embryonic development. Morpholino induced knockdown of both zebrafish ewsr1 genes led to mitotic defects with multipolar or otherwise abnormal mitotic spindles starting from the bud stage (10 hour post-fertilization (hpf)). The abnormalities in mitotic spindles were followed by p53-mediated apoptosis in the developing central nervous system (CNS) leading to a reduction in the number of proneural cells, disorganization of neuronal networks, and embryonic lethality by 5 days post-fertilization. siRNA silencing of EWSR1 in Hela cells resulted in mitotic defects accompanied by apoptotic cell death, indicating that the role of EWSR1 is conserved between zebrafish and human. Ewsr1 maintains mitotic integrity and proneural cell survival in early zebrafish development.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
MISSION® esiRNA, targeting human EWSR1