Merck
  • A novel radioresistant mechanism of galectin-1 mediated by H-Ras-dependent pathways in cervical cancer cells.
A novel radioresistant mechanism of galectin-1 mediated by H-Ras-dependent pathways in cervical cancer cells.
Cell death & disease (2012-01-13)
E-Y Huang, Y-F Chen, Y-M Chen, I-H Lin, C-C Wang, W-H Su, P-C Chuang, K-D Yang
ABSTRACT
Galectin-1 is a lectin recognized by galactoside-containing glycoproteins, and is involved in cancer progression and metastasis. The role of galectin-1 in radiosensitivity has not previously been investigated. Therefore, this study tests whether galectin-1 is involved in the radiosensitivity mediated by the H-Ras signaling pathway using cervical carcinoma cell lines. A knockdown of galectin-1 expression in HeLa cells decreased clonogenic survival following irradiation. The clonogenic survival increased in both HeLa and C33A cells with galectin-1 overexpression. The overexpression or knockdown of galectin-1 did not alter radiosensitivity, whereas H-Ras was silenced in both cell lines. Whereas K-Ras was knocked down, galectin-1 restored the radiosensitivity in HeLa cells and C33A cells. The knockdown of galectin-1 increased the high-dose radiation-induced cell death of HeLa cells transfected by constitutively active H-Ras. The knockdown of galectin-1 inhibited the radiation-induced phosphorylation of Raf-1 and ERK in HeLa cells. Overexpression of galectin-1 enhanced the phosphorylation of Raf-1 and ERK in C33A cells following irradiation. Galectin-1 decreased the DNA damage detected using comet assay and γ-H2AX in both cells following irradiation. These findings suggest that galectin-1 mediates radioresistance through the H-Ras-dependent pathway involved in DNA damage repair.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Millipore
Catch and Release v2.0 Reversible Immunoprecipitation System