Merck
  • Selective activation of cannabinoid CB(2) receptors suppresses spinal fos protein expression and pain behavior in a rat model of inflammation.
Selective activation of cannabinoid CB(2) receptors suppresses spinal fos protein expression and pain behavior in a rat model of inflammation.
Neuroscience (2003-06-18)
A G Nackley, A Makriyannis, A G Hohmann
ABSTRACT
Activation of cannabinoid CB(2) receptors attenuates thermal nociception in untreated animals while failing to produce centrally mediated effects such as hypothermia and catalepsy [Pain 93 (2001) 239]. The present study was conducted to test the hypothesis that activation of CB(2) in the periphery suppresses the development of inflammatory pain as well as inflammation-evoked neuronal activity at the level of the CNS. The CB(2)-selective cannabinoid agonist AM1241 (100, 330 micrograms/kg i.p.) suppressed the development of carrageenan-evoked thermal and mechanical hyperalgesia and allodynia. The AM1241-induced suppression of carrageenan-evoked behavioral sensitization was blocked by the CB(2) antagonist SR144528 but not by the CB(1) antagonist SR141716A. Intraplantar (ipl) administration of AM1241 (33 micrograms/kg ipl) suppressed hyperalgesia and allodynia following administration to the carrageenan-injected paw but was inactive following administration in the contralateral (noninflamed) paw, consistent with a local site of action. In immunocytochemical studies, AM1241 suppressed spinal Fos protein expression, a marker of neuronal activity, in the carrageenan model of inflammation. AM1241 suppressed carrageenan-evoked Fos protein expression in the superficial and neck region of the dorsal horn but not in the nucleus proprius or the ventral horn. The suppression of carrageenan-evoked Fos protein expression induced by AM1241 was blocked by coadministration of SR144528 in all spinal laminae. These data provide evidence that actions at cannabinoid CB(2) receptors are sufficient to suppress inflammation-evoked neuronal activity at rostral levels of processing in the spinal dorsal horn, consistent with the ability of AM1241 to normalize nociceptive thresholds and produce antinociception in inflammatory pain states.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
(R,S)-AM1241, ≥98% (HPLC), solid