Merck
The role of AEG-1/MTDH/LYRIC in the pathogenesis of central nervous system disease.
Advances in cancer research (2013-07-31)
Evan K Noch, Kamel Khalili
ABSTRACT
Astrocyte-elevated gene-1 (AEG-1/MTDH/LYRIC) is a potent oncogene that regulates key cellular processes underlying disease of the central nervous system (CNS). From its involvement in human immunodeficiency virus (HIV)-1 infection to its role in neurodegenerative disease and malignant brain tumors, AEG-1/MTDH/LYRIC facilitates cellular survival and proliferation through the control of a multitude of molecular signaling cascades. AEG-1/MTDH/LYRIC induction by HIV-1 and TNF highlights its importance in viral infection, and its incorporation into viral vesicles supports its potential role in active viral replication. Overexpression of AEG-1/MTDH/LYRIC in the brains of Huntington's disease patients suggests its function in neurodegenerative disease, and its association with genetic polymorphisms in large genome-wide association studies of migraine patients suggests a possible role in the pathogenesis of migraine headaches. In the field of cancer, AEG-1/MTDH/LYRIC promotes angiogenesis, migration, invasion, and enhanced tumor metabolism through key oncogenic signaling cascades. In response to external stress cues and cellular mechanisms to inhibit further growth, AEG-1/MTDH/LYRIC activates pathways that bypass cell checkpoints and potentiates signals to enhance survival and tumorigenesis. As an oncogene that promotes aberrant cellular processes within the CNS, AEG-1/MTDH/LYRIC represents an important therapeutic target for the treatment of neurological disease.