Merck
A polymorphic CA repeat marker at the human 27-kD calbindin (CALB1) locus.
Journal of human genetics (1999-11-26)
K Morishima, S Matsuura, H Tauchi, A Nakamura, K Komatsu
ABSTRACT
A polymorphic dinucleotide (CA) sequence was isolated from a BAC clone containing the human 27-kD calbindin (CALB1) gene at 8q21. This polymorphism will be a useful genetic marker to study genetic variations of the CALB1 gene.