Merck
  • Preparation and characterization of a halogen-bonded shape-persistent chiral alleno-acetylenic inclusion complex.
Preparation and characterization of a halogen-bonded shape-persistent chiral alleno-acetylenic inclusion complex.
Organic letters (2014-02-12)
Silvia Castro-Fernández, Inmaculada R Lahoz, Antonio L Llamas-Saiz, José Lorenzo Alonso-Gómez, María-Magdalena Cid, Armando Navarro-Vázquez
ABSTRACT
A chiral bidentate inclusion complex has been formed by halogen-bond interaction between the pyridyl moieties of a pyridoallenoacetylenic host and octafluorodiiodobutane. X-ray crystallography showed that the guest adopts a chiral conformation inside the molecular channels formed by stacking of the host units. A 10 ppm shielding of the (15)N NMR resonance for the pyridil units provided evidence of the formation of the halogen-bond complex in solution.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
Octafluoro-1,4-diiodobutane, 98%