Merck
  • A standardized method for the analysis of liver sinusoidal endothelial cells and their fenestrations by scanning electron microscopy.
A standardized method for the analysis of liver sinusoidal endothelial cells and their fenestrations by scanning electron microscopy.
Journal of visualized experiments : JoVE (2015-05-21)
Victoria C Cogger, Jennifer N O'Reilly, Alessandra Warren, David G Le Couteur
ABSTRACT
Liver sinusoidal endothelial cells are the gateway to the liver, their transcellular fenestrations allow the unimpeded transfer of small and dissolved substances from the blood into the liver parenchyma for metabolism and processing. Fenestrations are dynamic structures--both their size and/or number can be altered in response to various physiological states, drugs, and disease, making them an important target for modulation. An understanding of how LSEC morphology is influenced by various disease, toxic, and physiological states and how these changes impact on liver function requires accurate measurement of the size and number of fenestrations. In this paper, we describe scanning electron microscopy fixation and processing techniques used in our laboratory to ensure reproducible specimen preparation and accurate interpretation. The methods include perfusion fixation, secondary fixation and dehydration, preparation for the scanning electron microscope and analysis. Finally, we provide a step by step method for standardized image analysis which will benefit all researchers in the field.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
Molecular sieves, 13X, beads, 8-12 mesh