Merck
  • Genomic organization and structure of the 3' region of human MUC3: alternative splicing predicts membrane-bound and soluble forms of the mucin.
Genomic organization and structure of the 3' region of human MUC3: alternative splicing predicts membrane-bound and soluble forms of the mucin.
Biochemical and biophysical research communications (1999-10-08)
S C Crawley, J R Gum, J W Hicks, W S Pratt, J P Aubert, D M Swallow, Y S Kim
ABSTRACT
The MUC3 gene encodes a large, glycosylated mucin produced by intestinal epithelial cells to form a protective barrier against the external environment. Recently published cDNA sequences for the carboxyl-terminal region of MUC3 polypeptide indicated that rodent Muc3 possesses two epidermal growth factor (EGF)-like domains, and putative transmembrane and cytoplasmic domains, whereas the sequence of human MUC3 predicted termination after the first EGF-like domain. Here we describe the complete genomic sequence encompassing the carboxyl terminal region of human MUC3, revealing the boundaries of 11 exons. RT-PCR and cDNA library cloning experiments indicate that the gene is alternatively spliced, yielding a major membrane-bound form as well as multiple soluble forms. Thus, this work indicates that both membrane-bound and soluble MUC3 mucin proteins are produced by alternative splicing of a single gene. A potentially important polymorphism involving a Tyr residue with a proposed role in signalling is described as well.