- Recent contributions from the asymmetric aza-Michael reaction to alkaloids total synthesis.
Recent contributions from the asymmetric aza-Michael reaction to alkaloids total synthesis.
Natural product reports (2013-07-31)
Zacharias Amara, Joachim Caron, Delphine Joseph
PMID23896828
ABSTRACT
This review focuses on recent applications of the aza-Michael reaction in alkaloids total synthesis with a special emphasis on stereoselectivity. The report highlights achievements and challenges over the past five years and describes stereoselective intra- and inter-molecular conjugate addition of nitrogen-containing nucleophiles, including tandem and cascade processes. Total asymmetric syntheses of natural scaffolds, such as pyrrolidine, piperidine and "izidine" families, are depicted. Multi-step syntheses of highly challenging natural products are further detailed, assessing the scope of the stereocontrolled aza-Michael reaction as a powerful tool in alkaloid chemistry.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto