- Tuning of the critical temperature in iron(II) spin-crossover materials based on bridging polycyanidometallates: pentacyanidonitrosylferrate(II) and hexacyanidoplatinate(IV).
Tuning of the critical temperature in iron(II) spin-crossover materials based on bridging polycyanidometallates: pentacyanidonitrosylferrate(II) and hexacyanidoplatinate(IV).
Inorganic chemistry (2011-11-22)
Radovan Herchel, Zdeněk Trávníček, Radek Zbořil
PMID22098426
ABSTRACT
The reactions of iron(II) sulfate, 4-amino-3,5-di-2-pyridyl-4H-1,2,4-triazole (abpt), and pentacyanidonitrosylferrate(II) or hexacyanidoplatinate(IV) resulted in the formation of one-dimensional iron(II) spin-crossover compounds [Fe(abpt)(2)(μ-Fe(CN)(5)(NO))](n) (1) and [Fe(abpt)(2)(μ-Pt(CN)(6))](n) (2) with the spin-transition critical temperature near or above room temperature accompanied by thermochromism. Furthermore, it has been proven that the critical temperature T(c) is influenced by the type of dianionic polycyanidometallate within the series of discussed systems, and it changes in the sequence of [Fe(CN)(5)(NO)](2-) < [Pt(CN)(6))](2-) < [Ni(CN)(4))](2-) ≈ [Pd(CN)(4))](2-) ≈ [Pt(CN)(4))](2-).
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto