Merck
  • Facile coupling of propargylic, allylic and benzylic alcohols with allylsilane and alkynylsilane, and their deoxygenation with Et3SiH, catalyzed by Bi(OTf)3 in [BMIM][BF4] ionic liquid (IL), with recycling and reuse of the IL.
Facile coupling of propargylic, allylic and benzylic alcohols with allylsilane and alkynylsilane, and their deoxygenation with Et3SiH, catalyzed by Bi(OTf)3 in [BMIM][BF4] ionic liquid (IL), with recycling and reuse of the IL.
Organic & biomolecular chemistry (2012-08-11)
G G K S Narayana Kumar, Kenneth K Laali
ABSTRACT
Allyltrimethylsilane (allyl-TMS) reacts with propargylic alcohols 1a-1d in the presence of 10% Bi(OTf)(3) in [BMIM][BF(4)] solvent to furnish the corresponding 1,5-enynes in respectable isolated yields (87-93%) at room temperature. The utility of Bi(OTf)(3) as a superior catalyst was demonstrated in a survey study on coupling of allyl-TMS with employing several metallic triflates (Bi, Ln, Al, Yb) as well as, B(C(6)F(5))(3), Zn(NTf(2))(2) and Bi(NO(3))(3)·5H(2)O. Coupling of cyclopropyl substituted propargylic alcohol with allyl-TMS gave the skeletally intact 1,5-enyne and a ring opened derivative as a mixture. Coupling of propargylic/allylic alcohol with allyl-TMS resulted in allylation at both benzylic (2 isomers) and propargylic positions, as major and minor products respectively. The scope of this methodology for allylation of a series of allylic and benzylic alcohols was explored. Chemoselective reduction of a host of propargylic, propagylic/allylic, bis-allylic, allylic, and benzylic alcohols with Et(3)SiH was achieved in high yields with short reaction times. The same approach was successfully applied to couple representative propargylic and allylic alcohols with 1-phenyl-2-trimethylsilylacetylene. The recovery and reuse of the ionic liquid (IL) was gauged in a case study with minimal decrease in isolated yields after six cycles.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
1-Phenyl-2-trimethylsilylacetylene, 99%