Merck
  • Glial cell line-derived neurotrophic factor receptor alpha 2 (GFRA2) gene is associated with tardive dyskinesia.
Glial cell line-derived neurotrophic factor receptor alpha 2 (GFRA2) gene is associated with tardive dyskinesia.
Psychopharmacology (2010-04-07)
Renan P Souza, Vincenzo de Luca, Gary Remington, Jeffrey A Lieberman, Herbert Y Meltzer, James L Kennedy, Albert H C Wong
ABSTRACT
Tardive dyskinesia (TD) has a pharmacogenetic component in which the interaction of antipsychotic exposure with individual genetic variation mediates risk. The glial cell line-derived neurotrophic factor (GDNF) signalling pathway has been associated with neuroprotective effects in central dopaminergic neurons and spinal motor neurons. Clinical trials have also investigated whether GDNF may ameliorate Parkinson's disease symptoms. We tested whether variants in the GDNF receptor alpha 2 (GFRA2) gene could play a role in TD susceptibility evaluating 16 variants in 172 Caucasian schizophrenia subjects. We observed one significant allelic association (rs4739285, permuted p = 0.042) and two genotypic associations: rs4739285 under additive inheritance model and rs4739217 under dominant inheritance model (permuted p = 0.044). Moreover, carriers of the major alleles for both rs6587002 and rs4739217 presented significantly higher risk for TD (OR = 2.04, permuted p = 0.014), while subjects with the minor allele for rs4739217 and the major allele for rs6988470 were less likely to have TD (OR = 0.21, permuted p = 0.0007). Haplotype results indicate that the minor allele of the rs4739217 is a risk factor for TD (permuted allelic p = 0.074). Age was also a risk factor for TD in our sample (p = 0.0001). Taken together, our findings suggest that GFRA2 genetic variants and age may play a role in TD susceptibility, but further work is required to confirm these findings.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
Anti-GFRA2 antibody produced in rabbit, affinity isolated antibody
Prezzi e disponibilità al momento non sono disponibili